Learn More
Esculape Laboratoire™ Eyewash Bottles for First Aid
Product Code. 11891553
Click to view available options
Volume (Metric):
200 mL
500 mL
Composition:
Sterile Isotonic Solution
Sterile Ophthalmic Solution
Includes:
200 mL Blue Neutral pH Solution, 50 0mL Bottle Green Sterile Ophthalmic Solution
Cap
Cap, Storage Case, Belt Loop w/ Carrying Case
Cap, Storage, Carrying Case
Quantity:
10 Pk.
12 Pk.
Ea.
Unit Size:
Each
Pack of 10
Pack of 12
Product Code. 11891553
Brand: Esculape Laboratoire™ 8205255X10
Use Esculape™ Eyewash Bottles for First Aid has a carrying case with a belt loop for portabilitiy.
  • Storage case
  • Contain sterile isotonic solution
  • Ergonomic
Specifications
Product Type Eyewash Bottle
Composition Sterile Ophthalmic Solution
For Use With (Application) Portable Eyewash
Includes Cap
Quantity 10 Pk.
Sterility Sterile
Volume (Metric) 200 mL