Learn More
Knick Elektronische™ Portamess™ 904 Conductivity Meter
Click to view available options
Display Type:
LC
Unit Size:
Each
Description
- Smooth membrane keypad is durable and easy to clean. No sticking or crusty deposits.
- 15mm character height LC display with antireflection natural glass is scratchproof and resistant to chemicals
- Clear icons for operator guidance
- Integrated quiver protects the sensor and is replaceable. Transparent and easy to remove
- Blue ergonomic enclosure is resistant to impact and chemicals with watertight, diecast chassis (IP 66 protection)
- Cover protects against dirt and damage.
- Highly flexible lamellar joint can be folded back completely.
- Fold-out hook for hanging or standing up. Keeps both hands free for handling the sensor.
- Gold-plated, water-tight robust sockets
- Practical, adjustable carrying strap
Specifications
Specifications
| For Use With (Application) | Mobile conductivity measurement |
| Display Type | LC |
| Model | 904 |
| Keypad | Smooth membrane keypad |
| Height (Metric) | 160 mm |