Learn More
Knick Elektronische™ Portamess™ 913X Conductivity Meter
Product Code. 10015511
Click to view available options
Battery Type:
Alkaline AA (3)
Unit Size:
Each
Product Code. 10015511
Brand: Knick Elektronische™ 913XCOND
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Fit for digital communication. Knick Elektronische™ Portamess™ 913X Conductivity Meter is a clear, uncomplicated unit for mobile conductivity measurement.
  • Portamess™ 913 Cond portable Conductimeter for Atex Areas.
  • Remote-controllable and fit for digital communication.
  • Smooth membrane keypad is durable and easy to clean. No sticking or crusty deposits.
  • Large, easy to read 15mm character height LC display
  • Antireflection natural glass
  • Scratchproof and resistant to chemicals.
  • Clear icons for operator guidance.
  • Integraed quiver protects the sensor. Transparent. Easy to remove.
  • Blue, ergonomic enclosure is resistant to impact and chemicals. Water-tight diecast chassis. (IP 66 protection).
  • Cover protects against dirt and damage.
  • Highly flexible lamellar joint can be folded back completely.
  • Fold-out hook for hanging or standing up keeps both hands free for handling the sensor.
  • Robust, water-tight sockets are gold-plated.
  • Includes adjustable carrying strap.
Specifications
Includes Portamess™ 913 (X) Cond incl. batteries and sensor container; Carrying strap; User manual; Quickstart instructions in German, English and French; Interface cable with adapter for printer and PC; Paraly™ transfer software
For Use With (Application) Measures conductivity, salinity, TDS and temperature in industry, the environment, food processing and waste-water treatment
Certifications/Compliance The meter meets the EMC requirements of 89/336/EEC and the recommendations as per NAMUR NE 21. The meter is IP 66 protected to EN 60 529 (jet water from all directions).
Model Portamess™ 913 X Cond
Data Logging Records up to 100 measured values with the temperature, date and time
Battery Type Alkaline AA (3)
Battery Life approx. 1000 hrs.