Learn More
Knick Elektronische™ Portamess™ 913X Conductivity Meter
Click to view available options
Battery Type:
Alkaline AA (3)
Unit Size:
Each
Description
- Portamess™ 913 Cond portable Conductimeter for Atex Areas.
- Remote-controllable and fit for digital communication.
- Smooth membrane keypad is durable and easy to clean. No sticking or crusty deposits.
- Large, easy to read 15mm character height LC display
- Antireflection natural glass
- Scratchproof and resistant to chemicals.
- Clear icons for operator guidance.
- Integraed quiver protects the sensor. Transparent. Easy to remove.
- Blue, ergonomic enclosure is resistant to impact and chemicals. Water-tight diecast chassis. (IP 66 protection).
- Cover protects against dirt and damage.
- Highly flexible lamellar joint can be folded back completely.
- Fold-out hook for hanging or standing up keeps both hands free for handling the sensor.
- Robust, water-tight sockets are gold-plated.
- Includes adjustable carrying strap.
Specifications
Specifications
| Includes | Portamess™ 913 (X) Cond incl. batteries and sensor container; Carrying strap; User manual; Quickstart instructions in German, English and French; Interface cable with adapter for printer and PC; Paraly™ transfer software |
| For Use With (Application) | Measures conductivity, salinity, TDS and temperature in industry, the environment, food processing and waste-water treatment |
| Certifications/Compliance | The meter meets the EMC requirements of 89/336/EEC and the recommendations as per NAMUR NE 21. The meter is IP 66 protected to EN 60 529 (jet water from all directions). |
| Model | Portamess™ 913 X Cond |
| Data Logging | Records up to 100 measured values with the temperature, date and time |
| Battery Type | Alkaline AA (3) |
| Battery Life | approx. 1000 hrs. |