missing translation for 'onlineSavingsMsg'
Learn More

Potassium 4-carboxyphenyltrifluoroborate, 95%, Thermo Scientific™

Catalog Number p-7034221
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
850623-38-0
C7H5BF3KO2
228.019
MFCD04115737
NMZCPMQLYMSRAD-UHFFFAOYSA-N
potassium 4-carboxyphenyl trifluoroborate, potassium 4-carboxyphenyltrifluoroborate, potassium 4-carboxyphenyl trifluoroboranuide, potassium 4-carboxypheny trifluoroborate, pubchem11378, amtb110, potassium 4-carboxyphenyl-trifluoroboranuide, potassium ion 4-carboxyphenyl trifluoroboranuide, potassium 4-carboxyphenyl trifluoro borate 1-, potassium 4-carboxyphenyl-tris fluoranyl boranuide
23697341
potassium;(4-carboxyphenyl)-trifluoroboranuide
[B-](C1=CC=C(C=C1)C(=O)O)(F)(F)F.[K+]
This item is not returnable. View return policy
850623-38-0
C7H5BF3KO2
228.019
MFCD04115737
NMZCPMQLYMSRAD-UHFFFAOYSA-N
potassium 4-carboxyphenyl trifluoroborate, potassium 4-carboxyphenyltrifluoroborate, potassium 4-carboxyphenyl trifluoroboranuide, potassium 4-carboxypheny trifluoroborate, pubchem11378, amtb110, potassium 4-carboxyphenyl-trifluoroboranuide, potassium ion 4-carboxyphenyl trifluoroboranuide, potassium 4-carboxyphenyl trifluoro borate 1-, potassium 4-carboxyphenyl-tris fluoranyl boranuide
23697341
potassium;(4-carboxyphenyl)-trifluoroboranuide
[B-](C1=CC=C(C=C1)C(=O)O)(F)(F)F.[K+]

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
Formula Weight 228.02
Percent Purity 95%
Chemical Name or Material Potassium 4-carboxyphenyltrifluoroborate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'