missing translation for 'onlineSavingsMsg'
Learn More

Potassium Acetate, Certified AR for Analysis, Fisher Chemical™

Brand:  Fisher Chemical P/3761/53

Additional Details : CAS : 127-08-2 Poids : 0.50000kg

Packaging Plastic powder jar
Quantity 500g

 View more versions of this product

Product Code. 10599190

  • 54.00€ / 500g


Expédition estimée
09-12-2020
Add to basket

Description

Description

Specifications

Chemical Identifiers

127-08-2
98.142
SCVFZCLFOSHCOH-UHFFFAOYSA-M
517044
potassium;acetate
C2H3KO2
12458
potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920
CHEBI:32029
CC(=O)[O-].[K+]

Specifications

127-08-2
12458
SCVFZCLFOSHCOH-UHFFFAOYSA-M
potassium;acetate
517044
98.14g/mol
Plastic powder jar
292°C
500g
C2H3KO2
potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920
CC(=O)[O-].[K+]
98.142
CHEBI:32029
Solid
White
7.0 to 8.0
Videos
SDS Documents