missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium bicarbonate, Puriss., meets analytical spec. of BP, Ph. Eur., USP, E501, 99.5-101.0% (acidimetric), Honeywell Fluka™
Click to view available options
Quantity:
1 kg
25 kg
6 x 1 kg
Packaging:
Paper / Poly Bag
Poly bottle
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
CAS Max % | 100.0000% |
Quantity | 25 kg |
Assay Percent Range | 99.5 to 101.0% |
Linear Formula | KHCO3 |
Beilstein | 4535309 |
Formula Weight | 100.12 |
Grade | puriss. |
Packaging | Paper / Poly Bag |
Density | 2.17 g/cm3 |
Chemical Name or Material | Potassium bicarbonate, Puriss., meets analytical specification of BP, Ph. Eur., USP, E501, 99.5-101.0% (acidimetric) |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'