missing translation for 'onlineSavingsMsg'
Learn More

Potassium bis(oxalato)oxotitanate(IV) hydrate, Puratronic™, 99.997% (metals basis), Thermo Scientific Chemicals

Catalog Number p-7058574
Click to view available options
Quantity:
2 g
10 g
50 g
This item is not returnable. View return policy
14402-67-6
C4K2O9Ti
318.10
MFCD00150592
UHWHMHPXHWHWPX-UHFFFAOYSA-J
Potassium titanyloxolate
dipotassium oxotitaniumbis(ylium) dioxalate
[K+].[K+].O=[Ti++].[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O
This item is not returnable. View return policy
14402-67-6
C4K2O9Ti
318.10
MFCD00150592
UHWHMHPXHWHWPX-UHFFFAOYSA-J
Potassium titanyloxolate
dipotassium oxotitaniumbis(ylium) dioxalate
[K+].[K+].O=[Ti++].[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless
Odor Odorless
Linear Formula K2[TiO(C2O4)2]·xH2O
Quantity 2 g
UN Number UN3288
Formula Weight 318.10 (Anhydrous)
Percent Purity 99.997%
Grade Puratronic™
Physical Form Coarse Crystalline Powder
Assay (metals basis)
Chemical Name or Material Potassium bis (oxalato)oxotitanate (IV) hydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'