missing translation for 'onlineSavingsMsg'
Learn More

Potassium Bisnonafluoro-1-butanesulfonimidate, TRC

Product Code. 30137480
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137480 50 mg 50mg
30137475 10 mg 10mg
17992845 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137480 Supplier Toronto Research Chemicals Supplier No. TRCP69871850MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 129135-87-1
Formula Weight 618.8618
InChI Formula InChI=1S/C8F18NO4S2.K/c9-1(10,5(17,18)19)3(13,14)7(23,24)32(28,29)27-33(30,31)8(25,26)4(15,16)2(11,12)6(20,21)22;/q-1;+1
IUPAC Name potassium;bis(1,1,2,2,3,3,4,4,4-nonafluorobutylsulfonyl)azanide
Molecular Formula C8NO4F18S2K
Molecular Weight (g/mol) 619.28
SMILES [K+].FC(F)(F)C(F)(F)C(F)(F)C(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
Synonym 1,1,2,2,3,3,4,4,4-nonafluoro-N-[(nonafluorobutyl)sulfonyl]-1-butanesulfonamide Potassium Salt, Bis(nonafluorobutanesulfone)imide Potassium Salt, EF-N 442, Potassium Bis(nonafluorobutanesulfonimide), Potassium bis(nonafluorobutanesulfonyl)imide, Potassium bis(perfluorobutanesulfonyl)imide;
Chemical Name or Material Potassium Bisnonafluoro-1-butanesulfonimidate

Specifications

Quantity 50 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.