missing translation for 'onlineSavingsMsg'
Learn More

Potassium Bisulfate, Reagent grade, Honeywell™

Catalog Number p-7120680
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Poly Bottle
This item is not returnable. View return policy
7646-93-7
HKO4S
136.16
MFCD00011404
CHKVPAROMQMJNQ-UHFFFAOYSA-M
potassium bisulfate, potassium hydrogen sulfate, potassium bisulphate, sulfuric acid, monopotassium salt, potassium acid sulfate, sal enixum, potassium hydrogensulphate, monopotassium sulfate, acid potassium sulfate, caswell no. 682c
516920
potassium;hydrogen sulfate
[K+].OS([O-])(=O)=O
This item is not returnable. View return policy
7646-93-7
HKO4S
136.16
MFCD00011404
CHKVPAROMQMJNQ-UHFFFAOYSA-M
potassium bisulfate, potassium hydrogen sulfate, potassium bisulphate, sulfuric acid, monopotassium salt, potassium acid sulfate, sal enixum, potassium hydrogensulphate, monopotassium sulfate, acid potassium sulfate, caswell no. 682c
516920
potassium;hydrogen sulfate
[K+].OS([O-])(=O)=O

Reagent Grade

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY
Quantity 500 g
Linear Formula KHSO4
UN Number UN2509
Formula Weight 136.17g/mol
Grade Reagent
Packaging Poly Bottle
Density 2.32g/mL at 25°C
Chemical Name or Material Potassium Bisulfate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'