missing translation for 'onlineSavingsMsg'
Learn More

Potassium bromide, Premium FTIR Grade, Thermo Scientific™

Brand:  Thermo Scientific Alfa Aesar 047111.KM

Product Code. 15432847

  • 162.00€ / Pack of 25
 Restricted Item
In Stock
This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

7758-02-3
BrK
MFCD00011358
potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095
[K+].[Br-]
119.00
CHEBI:32030
Premium FTIR
0.5g packets
2.75 g/mL
25 pc.
KBr
14,7618
IOLCXVTUBQKXJR-UHFFFAOYSA-M
potassium;bromide
253877
119.01
Odorless
1.559
Potassium bromide
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.