missing translation for 'onlineSavingsMsg'
Learn More

Potassium chloride, 0.0375M, Conductivity Standard, Thermo Scientific Chemicals

Catalog Number p-7052160
Click to view available options
Quantity:
500 mL
1 L
4 L
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
7447-40-7
ClK
74.55
MFCD00011360
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
4873
CHEBI:32588
potassium chloride
[Cl-].[K+]
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
7447-40-7
ClK
74.55
MFCD00011360
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
4873
CHEBI:32588
potassium chloride
[Cl-].[K+]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Concentration 0.0375M
Quantity 500 mL
Formula Weight 74.55
Odor Odorless
Physical Form Liquid
Chemical Name or Material Potassium chloride Conductivity Standard, 5000μmho at 25°C

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'