missing translation for 'onlineSavingsMsg'
Learn More

Potassium Chloride Saturated Solution, Pure, Electrode Filling Solution, Fisher Chemical™

Product Code. 10744181 Shop All Fisher Chemical Products
Click to view available options
Quantity:
1 L
100 mL
250 mL
Packaging:
Amber Glass Bottle
HDPE Plastic Bottle
Unit Size:
0.25 L
1 L
100mL
This item is not returnable. View return policy

Product Code. 10744181

Brand: Fisher Chemical J/2983/05

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 7447-40-7, 7732-18-5
Molecular Formula ClK
Molecular Weight (g/mol) 74.55
MDL Number MFCD00011360
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
PubChem CID 4873
ChEBI CHEBI:32588
SMILES [Cl-].[K+]

Specifications

Boiling Point 106°C
Physical Form Liquid
Quantity 100 mL
Synonym potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs
IUPAC Name potassium;chloride
Packaging Amber Glass Bottle
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.