missing translation for 'onlineSavingsMsg'
Learn More

Potassium Cyanide, puriss. p.a. ACS Reagent, Honeywell Fluka™

Catalog Number p-7072011
Click to view available options
Quantity:
100 g
250 g
1 kg
25 kg
This item is not returnable. View return policy
151-50-8
CKN
65.116
MFCD00011397
NNFCIKHAZHQZJG-UHFFFAOYSA-N
potassium cyanide, potassium cyanide k cn, cyanide of potassium, caswell no. 688a, zyankali, kalium cyanid german, unii-mqd255m2zo, rcra waste number p098, cyanure de potassium french
9032
potassium;cyanide
[C-]#N.[K+]
This item is not returnable. View return policy
151-50-8
CKN
65.116
MFCD00011397
NNFCIKHAZHQZJG-UHFFFAOYSA-N
potassium cyanide, potassium cyanide k cn, cyanide of potassium, caswell no. 688a, zyankali, kalium cyanid german, unii-mqd255m2zo, rcra waste number p098, cyanure de potassium french
9032
potassium;cyanide
[C-]#N.[K+]

Puriss. p.a., ACS Reagent, Reag. Ph. Eur.,≥97.0%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Quantity 100 g
Linear Formula KCN
UN Number 1680
Beilstein 3593645
Formula Weight 65.12g/mol
Percent Purity ≥97%
Grade ACS/puriss. p.a.
Chemical Name or Material Potassium Cyanide, Reag. Ph. Eur.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'