missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium dichromate concentrate, For 1L standard solution, 1/60 M K2Cr2O7 (0.1N), Honeywell Fluka™
Click to view available options
Quantity:
1 Pc.
6 x 1 Ea.
Packaging:
Ampoule
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Quantity | 1 Pc. |
Linear Formula | K2Cr2O7 |
UN Number | UN3082 |
Formula Weight | 294.18g/mol |
Packaging | Ampoule |
Chemical Name or Material | Potassium dichromate concentrate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'