missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Hydrogen Diiodate, ≥99.8% (RT), Honeywell Fluka™
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Color | White |
Physical Form | Powder |
Quantity | 500 g |
Linear Formula | KH(IO3)2 |
UN Number | UN3085 |
Formula Weight | 389.91g/mol |
Percent Purity | ≥99.8% (RT) |
Packaging | Glass bottle |
Chemical Name or Material | Potassium hydrogen diiodate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'