Learn More
Potassium Iodate, ACS Reagent, 99.5%, Honeywell™ Fluka™
ACS Reagent, 99.5%
Brand: Honeywell Fluka 215929-2.5KG
Additional Details : CAS : 7758-05-6 Poids : 2.50000kg Transport : UN number : 1479 Chem class : 5.1 Pack group : II
Packaging | Poly Bottle |
---|---|
Quantity | 2.5kg |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
7758-05-6 | |
214.00 | |
JLKDVMWYMMLWTI-UHFFFAOYSA-M | |
23665710 | |
[O-]I(=O)=O.[K+] |
IKO3 | |
MFCD00011406 | |
potassium iodate, potassium triodate, caswell no. 693a, iodic acid, potassium salt, unii-i139e44nhl, potassium iodine oxide kio3, iodic acid hio3 , potassium salt, epa pesticide chemical code 075703, iodic acid hio3 , potassium salt 1:1, kaliumjodat | |
potassium;iodate |
Specifications
Potassium Iodate | |
IKO3 | |
MFCD00011406 | |
Crystals | |
JLKDVMWYMMLWTI-UHFFFAOYSA-M | |
potassium;iodate | |
23665710 | |
99.50% | |
ca. 2,400kg/cu.m | |
Poly Bottle | |
Colorless | |
5 to 8 (at 20°C) |
7758-05-6 | |
KIO3 | |
UN1479 | |
potassium iodate, potassium triodate, caswell no. 693a, iodic acid, potassium salt, unii-i139e44nhl, potassium iodine oxide kio3, iodic acid hio3 , potassium salt, epa pesticide chemical code 075703, iodic acid hio3 , potassium salt 1:1, kaliumjodat | |
[O-]I(=O)=O.[K+] | |
214.00 | |
214g/mol | |
ACS Reagent | |
3.93g/mL at 25°C | |
Solid | |
560°C | |
2.5kg |