missing translation for 'onlineSavingsMsg'
Learn More

Potassium molybdate, 95%, Thermo Scientific Chemicals

Catalog Number p-4440153
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
13446-49-6
K2MoO4
238.14
MFCD00011370
NUYGZETUIUXJCO-UHFFFAOYSA-N
potassium molybdenum oxide, dipotassium dioxido dioxo molybdenum, dipotassium ion molybdate, 2k.moo4, potassium molybdenum oxide, anhydrous
6093816
dipotassium;dioxido(dioxo)molybdenum
[K+].[K+].[O-][Mo]([O-])(=O)=O
This item is not returnable. View return policy
13446-49-6
K2MoO4
238.14
MFCD00011370
NUYGZETUIUXJCO-UHFFFAOYSA-N
potassium molybdenum oxide, dipotassium dioxido dioxo molybdenum, dipotassium ion molybdate, 2k.moo4, potassium molybdenum oxide, anhydrous
6093816
dipotassium;dioxido(dioxo)molybdenum
[K+].[K+].[O-][Mo]([O-])(=O)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 919.0°C
Quantity 100 g
Assay Percent Range 94% min
Linear Formula K2MoO4
Merck Index 15,7768
Formula Weight 238.14
Percent Purity 95%
Packaging Glass bottle
Chemical Name or Material Potassium molybdate
compliance-icons
Product Identifier
  • Potassium molybdate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'