missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium phosphate monobasic, ACS Reagent, Honeywell Fluka™
Click to view available options
Quantity:
25 g
500 g
1 kg
2.5 kg
6 x 500 g
4 x 2.5 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Quantity | 25 g |
Linear Formula | KH2PO4 |
UN Number | NONH for all modes of transport |
Formula Weight | 136.09g/mol |
Percent Purity | ≥99% |
Grade | ACS Reagent |
Density | 2.338g/mL at 25°C |
Chemical Name or Material | Potassium phosphate monobasic |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'