missing translation for 'onlineSavingsMsg'
Learn More

Potassium Sodium Tartrate Tetrahydrate, Extra Pure, SLR, Fisher Chemical™

Catalog Number p-7073448
Click to view available options
Quantity:
500 g
1 kg
5 kg
Packaging:
Plastic keg
Plastic powder jar
This item is not returnable. View return policy
6381-59-5
C4H12KNaO10
282.218
150989
VZOPRCCTKLAGPN-UHFFFAOYSA-L
potassium sodium tartrate tetrahydrate, sodium potassium tartrate tetrahydrate, monopotassium monosodium tartrate tetrahydrate, sodiumpotassiumtartrate,tetr, potassium sodium 2,3-dihydroxybutanedioate tetrahydrate, potassium sodium tetrahydrate tartrate, l +-tartaric acid potassium sodium salt, acmc-209nif, acmc-20aju8, ksc493o7j
2724148
potassium;sodium;2,3-dihydroxybutanedioate;tetrahydrate
C(C(C(=O)[O-])O)(C(=O)[O-])O.O.O.O.O.[Na+].[K+]
This item is not returnable. View return policy
6381-59-5
C4H12KNaO10
282.218
150989
VZOPRCCTKLAGPN-UHFFFAOYSA-L
potassium sodium tartrate tetrahydrate, sodium potassium tartrate tetrahydrate, monopotassium monosodium tartrate tetrahydrate, sodiumpotassiumtartrate,tetr, potassium sodium 2,3-dihydroxybutanedioate tetrahydrate, potassium sodium tetrahydrate tartrate, l +-tartaric acid potassium sodium salt, acmc-209nif, acmc-20aju8, ksc493o7j
2724148
potassium;sodium;2,3-dihydroxybutanedioate;tetrahydrate
C(C(C(=O)[O-])O)(C(=O)[O-])O.O.O.O.O.[Na+].[K+]
Melting Point 70°C
Color White
pH 5.5 to 8.5
Physical Form Powder/Solid
Quantity 500 g
Formula Weight 282.22g/mol
Packaging Plastic powder jar
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'