Learn More
Thermo Scientific Chemicals Potassium sulfate, 99+%, for biochemistry
Catalog Number p-43506
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Plastic Bottle
7778-80-5
K2O4S
174.25
MFCD00011388
OTYBMLCTZGSZBG-UHFFFAOYSA-L
potassium sulfate, dipotassium sulfate, potassium sulphate, sulfuric acid dipotassium salt, sal polychrestum, sulfuric acid, potassium salt, arcanum duplicatum, kalium sulphuricum, caswell no. 702, sulfuric acid, dipotassium salt
24507
CHEBI:32036
dipotassium sulfate
[K+].[K+].[O-]S([O-])(=O)=O
7778-80-5
K2O4S
174.25
MFCD00011388
OTYBMLCTZGSZBG-UHFFFAOYSA-L
potassium sulfate, dipotassium sulfate, potassium sulphate, sulfuric acid dipotassium salt, sal polychrestum, sulfuric acid, potassium salt, arcanum duplicatum, kalium sulphuricum, caswell no. 702, sulfuric acid, dipotassium salt
24507
CHEBI:32036
dipotassium sulfate
[K+].[K+].[O-]S([O-])(=O)=O
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Boiling Point 1689.0°C
Melting Point 1067.0°C
Color White
Physical Form Crystals
Quantity 250 g
Assay Percent Range 99+%
Linear Formula K2SO4
Merck Index 15,7793
Solubility Information (10 g in 150 ml water) Clear colorless
Formula Weight 174.25
Percent Purity ≥99%
Grade Biochemical
Packaging Plastic Bottle
Chemical Name or Material Potassium sulfate