Learn More
Potassium Tetraiodomercurate Solution A, Ph. Eur. Grade, Reagecon™
BIOPHARMA
Product Code. 12524376 Shop All Reagecon Products
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
Product Code. 12524376
Brand: Reagecon 1071600A
Specifications
Quantity 100 mL
compliance-icons compliance-icons compliance-icons
Product Identifier
  • POTASSIUM TETRAIODOMERCURATE SOLUTION - A
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 1
  • Specific target organ toxicity after repeated exposure Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H301-Toxic if swallowed.
  • H310-Fatal in contact with skin.
  • H331-Toxic if inhaled.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P330-Rinse mouth.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : mercury diiodide