missing translation for 'onlineSavingsMsg'
Learn More

Witeg™ Powder Funnel

Funnel powders quickly using the short and wide stem of this funnel. Witeg™ Powder Funnel, constructed of resistant and durable Duran and is designed with a 60° angle and a 50mm O.D.

Brand:  Witeg™ 5 790 001

Additional Details : Weight : 0.80000kg

Product Code. 11897830

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.


Description

Description

  • Short and wide stem.
  • Duran construction.
  • O.D.: 50mm.
  • Angle: 60°.
Specifications

Specifications

50mm
Glass
60°
Short/wide stem
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions