Learn More
Aalborg™ Power Supply
Click to view available options
For Use With (Equipment):
Digital Flowmeter
Gas Mass Flow Controller
Gas Mass Flowmeter
Type:
DFM Adapter
GFC Adapter
Power Supply
Unit Size:
Each
Description
- Accessory for DFM, GFM, or GFC
- 230VAC Continential Europe Plug 12VDC
- Power supply
Specifications
Specifications
| Voltage | 230 V |
| For Use With (Equipment) | Digital Flowmeter |
| Type | DFM Adapter |
| Electrical Requirements | 230VAC, 12VDC |