missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ PP 242

Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
Description
PP 242 is an ATP-competitive mTORC1/mTORC2 inhibitor (IC50 = 8 nM). Displays selectivity for mTOR over other PI 3K family kinases (IC50 values are 0.102, 0.408, 1.27, 1.96 and 2.2 μM for p110γ, DNA-PK, p110δ, p110α and p110β respectively) and 215 further kinases. Displays modest inhibition of PKCα, JAK2, PKCβI, PKCβII and RET (IC50 values are 0.049, 0.110, 0.185, 0.198 and 0.224 μM respectively). Inhibits both S6K and 4EBP1 phosphorylation; activity causes a decrease in cap-dependent protein translation. Also triggers downregulation of cFLIPS and augments TRAIL-induced apoptosis of cancer cells. PP 242 also inhibits oncogenic K-Ras and PI 3-K induced lipogenesis in breast cancer cells.
Specifications
Specifications
| Chemical Name or Material | 2-[4-Amino-1-(1-methylethyl)-1H-pyrazolo[3,4-d]pyrimidin-3-yl]-1H-indol-5-ol |
| CAS | 1092351-67-1 |
| Assay Percent Range | >98% |
| Quantity | 5 mg |
| Molecular Formula | C16H16N6O |
| Formula Weight | Observed MW: 308.34 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction