missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ PPADS tetrasodium salt
GREENER_CHOICE

Product Code. 18024922
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18024922

Brand: Tocris Bioscience™ 0625/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Non-selective P2 antagonist

PPADS tetrasodium salt is a non-selective P2 purinergic antagonist. Blocks recombinant P2X1, P2X2, P2X3, P2X5 (IC50 = 1 - 2.6 μM), native P2Y2-like (IC50 ∼ 0.9 mM), and recombinant P2Y4 (IC50 ∼ 15 mM) receptors. PPADS tetrasodium salt delays onset of calcium responses to mild hypoosmotic stress in cortical slices. PPADS tetrasodium salt blocks P2X7 and reduces clinical and histological graft-versus-host disease development in a humanised mouse model.

2'5'-isomer also available.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 192575-19-2
Molecular Formula C14H10N3Na4O12PS2

Specifications

Quantity 50 mg
Formula Weight 599.3
Purity >98%
Molecular Weight (g/mol) 599.296
Chemical Name or Material PPADS tetrasodium salt
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.