missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ PPQ 102
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
PPQ 102 is a potent, voltage-independent CFTR chloride channel inhibitor (IC50 = 90 nM). Reduces the size and number of renal cysts in an embryonic kidney culture model of polycystic kidney disease. Also increases VEGF-A production from airway epithelial cells in vitro.
Specifications
Specifications
| Purity | >98% |
| Quantity | 10 mg |
| Product Type | PPQ 102 |
| Molecular Weight (g/mol) | 438.48 |
| Inhibitors | PPQ 102 |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction