missing translation for 'onlineSavingsMsg'
Learn More

Praseodymium rod, 12.7mm (0.5in) dia, 99.5% (metals basis excluding Ta), Thermo Scientific Chemicals

Catalog Number p-7051022
Click to view available options
Quantity:
10 mm
25 mm
50 mm
100 mm
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
7440-10-0
Pr
140.91
MFCD00011174
PUDIUYLPXJFUGB-UHFFFAOYSA-N
unii-nkn7eza750, nkn7eza750, ingot, praseodimio, praseodyme, praseodym, atom, chips, powder, acmc-1bktg
23942
CHEBI:49828
praseodymium
[Pr]
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
7440-10-0
Pr
140.91
MFCD00011174
PUDIUYLPXJFUGB-UHFFFAOYSA-N
unii-nkn7eza750, nkn7eza750, ingot, praseodimio, praseodyme, praseodym, atom, chips, powder, acmc-1bktg
23942
CHEBI:49828
praseodymium
[Pr]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
TRUSTED_SUSTAINABILITY
Quantity 10 mm
Percent Purity 99.5%
Odor Odorless
Packaging Mylar under Argon
Weight ≈0.86g/mm
Form Cast Rod
Assay (metals basis excluding Ta)
Chemical Name or Material Praseodymium rod, 12.7mm (0.5 in.) dia.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'