Learn More
Tocris Bioscience™ Prasugrel
GREENER_CHOICE
Irreversible P2Y12 antagonist; antiplatelet and orally active
Brand:  Tocris Bioscience™ 6317/50
Encompass
Product Code. 15736409
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Prasugrel is a irreversible P2Y12 antagonist; metabolised to active metabolite R-99224 (IC50 = 45 μM) irreversibly binds platelet P2Y12 receptors, inhibiting platelet activation. Antiplatelet and antithrombotic. Orally active and active in vivo. Note - inactive in vitro.
Specifications
Specifications
2-[2-(Acetyloxy)-6,7-dihydrothieno[3,2-c]pyridin-5(4H)-yl]-1-cyclopropyl-2-(2-fluorophenyl)ethanone
150322-43-3
>98%