Learn More
Tocris Bioscience™ Prasugrel
Irreversible P2Y12 antagonist; antiplatelet and orally active
Brand:  Tocris Bioscience™ 6317/10
Encompass
Product Code. 15726409
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Description
Description
Prasugrel is a irreversible P2Y12 antagonist; metabolised to active metabolite R-99224 (IC50 = 45 μM) irreversibly binds platelet P2Y12 receptors, inhibiting platelet activation. Antiplatelet and antithrombotic. Orally active and active in vivo. Note - inactive in vitro.
Specifications
Specifications
2-[2-(Acetyloxy)-6,7-dihydrothieno[3,2-c]pyridin-5(4H)-yl]-1-cyclopropyl-2-(2-fluorophenyl)ethanone
150322-43-3
>98%