missing translation for 'onlineSavingsMsg'
Learn More

Prazosin hydrochloride, Thermo Scientific™

An alpha adrenoceptor antagonist

Brand:  Thermo Scientific Alfa Aesar J61712.03

Quantity 1g

 View more versions of this product

Product Code. 15474249

  • 251.00€ / 1g
 Restricted Item
Estimated Shipment: 25-09-2026
This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Prazosin hydrochloride is an inhibitor of PDE. Also offered in a free base form Prazosin

Solubility
Soluble in dimethyl sulfoxide, ethanol, water, methanol.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

19237-84-4
419.87
WFXFYZULCQKPIP-UHFFFAOYSA-N
68546
hydrogen 2-[4-(furan-2-carbonyl)piperazin-1-yl]-6,7-dimethoxyquinazolin-4-amine chloride
C19H22ClN5O4
MFCD00058177
prazosin hydrochloride, prazosin hcl, minipress, vasoflex, peripress, furazosin hydrochloride, deprazolin, hypovase, hypovasole, pratsiol
CHEBI:8365
[H+].[Cl-].COC1=C(OC)C=C2C(N)=NC(=NC2=C1)N1CCN(CC1)C(=O)C1=CC=CO1

Specifications

19237-84-4
MFCD00058177
4303561
prazosin hydrochloride, prazosin hcl, minipress, vasoflex, peripress, furazosin hydrochloride, deprazolin, hypovase, hypovasole, pratsiol
WFXFYZULCQKPIP-UHFFFAOYSA-N
hydrogen 2-[4-(furan-2-carbonyl)piperazin-1-yl]-6,7-dimethoxyquinazolin-4-amine chloride
68546
419.86
C19H22ClN5O4
1g
14,7717
Soluble in dimethyl sulfoxide,ethanol,water,methanol.
[H+].[Cl-].COC1=C(OC)C=C2C(N)=NC(=NC2=C1)N1CCN(CC1)C(=O)C1=CC=CO1
419.87
CHEBI:8365
Prazosin hydrochloride
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.