Learn More
Lauda™ Pre-Filter
Pre-Filter with fine filter candle.
Brand:  Lauda™ A000133
Additional Details : Weight : 1.00000kg
Product Code. 16697485
  • 459.00€ / Each
Estimated Shipment: 18-06-2024
to see stock.
Add to basket
Description
Description
  • For use with all Puridest Water Stills
Specifications
Specifications
1 μm
Fine Filter Candle