Learn More
Fisherbrand™ Precision Hydrometer Acc. to ASTM E100-10
Product Code. 11716943
Encompass
Click to view available options
Density (Metric):
1000 to 1050g/L
600 - 650 kg/m3
650 - 700 kg/m3
700 - 750 kg/m3
750 - 800 kg/m3
800 - 850 kg/m3
850 - 900 kg/m3
900 to 950g/L
950 - 1000 kg/m3
ASTM Number:
300H
301H
302H
303H
304H
305H
306H
307H
308H
Unit Size:
Each
Product Code. 11716943
Brand: Fisherbrand™ 2304H
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Combined form glass density hydrometers with mercury thermometers in the body. Fisherbrand™ Precision Hydrometer Acc. to ASTM E100-10 measure in kg/m3 and operate in a wide temperature range of -20 to 65°C, with a reference temperature of 15°C.
High-precision instruments for determination of fluid density in accordance with ASTM standards and available with a DAkkS/works calibration certificate.
  • Meets the standards of ASTM E100-10 for density hydrometers
  • Reference temperature: 15°C
  • Scale division: 0.5kg/m3
  • Error limit: ±0.5kg/m3
  • Mercury Thermometer: -20 to +65°C in 1°C
  • Total length: 380mm
  • Single packed in transparent square plastic sleeve
Specifications
Type Hydrometer with Incorporated Thermometer
ASTM Number 304H
Accuracy ±0.5 kg/m3
Increments Density: 0.5 kg/m3, Temperature: 1°C
Certifications/Compliance ASTM E100-10
Length (Metric) 380 mm
Includes Hg Thermometer in body, Square plastic sleeve
Density (Metric) 800 - 850 kg/m3
Temperature Accuracy ±1° C
Material Glass
Temperature Range (Metric) −20 to +65°C
For Use With (Application) Determination of fluid density
Temperature of Standardization 15°C