Learn More
Fisherbrand™ Precision Hydrometers for Specific Gravity
Click to view available options
Specific Gravity Range:
0.590 to 0.660
1.200 to 1.420
1.400 to 1.620
1.600 to 1.670
1.660 to 1.730
Increments:
0.0005 sp.gr.
0.002 sp.gr.
Length (Metric):
300 mm
325 mm
Unit Size:
Each
Description
High-precision instruments for determination of fluid density available with a DAkkS or works calibration certificate.
- Reference temperature: 60/60°F
- Without thermometer – control thermometer sold separately
- Single packed in transparent square plastic sleeve
Specifications
Specifications
| Type | Hydrometer without Thermometer |
| Accuracy | ±0.002 sp/gr |
| Increments | 0.002 sp.gr. |
| Length (Metric) | 300 mm |
| For Use With (Equipment) | Control thermometer |
| Includes | Plastic sleeve |
| Material | Glass |
| For Use With (Application) | Measuring specific gravity |
| Specific Gravity Range | 1.400 to 1.620 |
| Temperature of Standardization | 60°F |