Learn More
SPEX™ Prep-Aid Vertrel XF
Product Code. 15330542 Shop All SPEX Certiprep Products
Click to view available options
:
Product Type: Grinding Aid
Product Code. 15330542
Brand: SPEX™ 3650
This item requires a price update prior to being ordered.
Request a Price for this item here.
SPEX™ Prep-Aid Vertrel XF is a liquid fluorocarbon grinding aid that prevents caking, reduces contamination, and evaporates after grinding.
  • Prevents caking and reduces contamination
  • Quickly evaporates after grinding without leaving any residue
  • Enhances the grindability of almost any sample
  • Makes the grinding container easier to clean
  • Particularly useful for grinding samples such as cement, rock, and clinker to particle sizes below 10 microns
Specifications
Product Type Grinding Aid
For Use With (Application) Grinding samples
Packaging Quantity 946.35 mL bottle