missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ preQ1-biotin

Description
preQ1-biotin is a modified analog of the natural substrate prequeosine1 (preQ1) and is used for RNA-TAG (transglycosylation at guanosine) and DNA-TAG. preQ1-biotin enables affinity tagging and pull-down of specific RNAs that have been modified selectively by E. coli tRNA guanine transglycosylase (TGT). preQ1-biotin is incorporated site-specifically and covalently into RNAs containing a short harpin nucleotide recognition motif. The guanine in a UGU recognition element is exchanged with the preQ1-biotin substrate.
Plasmids for expression of the E.coli TGT enzyme ( #138201 ) and for cloning an RNA of interest into a vector containing the recognition motif at the 3' end ( #138209 ) are available from Addgene.
Specifications
Specifications
| Chemical Name or Material | N-(6-(((2-Amino-4-oxo-4,7-dihydro-3H-pyrrolo[2,3-d]pyrimidin-5-yl)methyl)amino)hexyl)-5-((3aS,4S,6aR)-2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanamide |
| Quantity | 5 mg |
| Target | Buffers, Chelators and Reagents |
| Molecular Formula | C23H36N8O3S |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?