Learn More
Thermo Scientific™ Pretreatment Cartridges and Filters
Choose one of these Thermo Scientific™ Pretreatment Cartridges and Filters when the quality of your feed water doesn't match the requirements for your point of use water purification system.
Brand: Thermo Scientific™ 06.5204
Additional Details : Weight : 1.00000kg
Description
Use Thermo Scientific™ Pretreatment Cartridges and Filters when the quality of your feed water doesn't match the requirements for your Thermo Scientific™ Barnstead™ water purification system.
Use pretreatment when your feed water quality doesn't match the requirements for your system.Specifications
5µm filter with hardness stabilizer | |
LabTower EDI, LabTower RO | |
Filter with Stabilizer |