Learn More
Heidolph™ Printed Circuit Board
Shop All Heidolph Instruments Products
Click to view available options
Voltage:
230V
Frequency:
50 Hz
For Use With (Equipment):
Hri-Smg 2000
Unit Size:
Each
Specifications
Specifications
| For Use With (Equipment) | Hri-Smg 2000 |
| Frequency | 50 Hz |
| Type | Printed Circuit Board |
| Voltage | 230V |