missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human PRKACB (PRKAC beta), GST Tag Recombinant Protein

Human PRKACB (PRKAC beta), GST Tag Recombinant Protein

Brand:  Invitrogen™ PV6390

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15409023

  • 7490.00€ / 1mg
Estimated Shipment: 14-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The PKA catalytic subunit consists of three gene products: C-alpha, C-beta, and C-gamma. The catalytic subunit C-beta has been assigned to human chromosome region 1p36.1. PRKACB is derived from a gene distinct from C-alpha and shows tissue-specific expression. At the amino acid level C-alpha and C-beta showed 93% homology.

  • Validated in LanthaScreen™ Kinase Binding Assay
  • Protocols available for simple use and optimized performance
  • Multiple pack sizes available for convenient use
Specifications

Specifications

Enzyme and Protein Activity Assay, Industrial and Applied Science, Kinase Assay, Kinase Biology, Kinase Proteins and Expression Reagents, Pharma and Biopharma, Protein Assays and Analysis, Protein Biology, Target and Lead Identification and Validation
66.9kDa
Store at -68 to -85°C. Aliquot at first use to avoid multiple freeze-thaws.
Human
GST Tag
50mM Tris-HCl pH 7.5, 150mM NaCl, 0.1mM EGTA, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol
1mg
PKAb, cAPKb, MGC9320, MGC41879, DKFZp781I2452
Recombinant
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions