missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Proadifen hydrochloride

Brand:  Alfa Aesar™ J63833.04

Additional Details : CAS : 62-68-0 Weight : 0.00200kg

Quantity 2g

 View more versions of this product

Product Code. 15405999

  • 293.00€ / 2g


Estimated Shipment
12-05-2021
Add to basket

Description

Description

Applications
Proadifen hydrochloride is an inhibitor of NOS1, AChR, cytochrome P-450, and KIR6. Proadifen hydrochloride bind to the protein moiety of P-450, also reduced the hypoxic response. of Proadifen hydrochloride have good antiarrhythmic effects. SKF-525A substantially reduced the depletion of cardiac norepinephrine.

Solubility
Soluble in methanol or water

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, heat.
Specifications

Chemical Identifiers

62-68-0
389.964
FHIKZROVIDCMJA-UHFFFAOYSA-N
65341
CCCC(C1=CC=CC=C1)(C2=CC=CC=C2)C(=O)OCCN(CC)CC.Cl
C23H32ClNO2
MFCD00055151
proadifen hydrochloride, proadifen hcl, propyladiphenin, skf-525a hydrochloride, skf 525a, sk&f 525-a, proadifen hydrochloride usan, skf 525-a, skf-525a, hydrochloride
2-(diethylamino)ethyl 2,2-diphenylpentanoate;hydrochloride

Specifications

Proadifen hydrochloride
122°C to 123°C
62-68-0
MFCD00055151
Soluble in methanol or water
CCCC(C1=CC=CC=C1)(C2=CC=CC=C2)C(=O)OCCN(CC)CC.Cl
389.964
389.97
White
2g
C23H32ClNO2
proadifen hydrochloride, proadifen hcl, propyladiphenin, skf-525a hydrochloride, skf 525a, sk&f 525-a, proadifen hydrochloride usan, skf 525-a, skf-525a, hydrochloride
FHIKZROVIDCMJA-UHFFFAOYSA-N
2-(diethylamino)ethyl 2,2-diphenylpentanoate;hydrochloride
65341
Powder
Videos
SDS Documents