missing translation for 'onlineSavingsMsg'
Learn More

Proadifen hydrochloride, Thermo Scientific Chemicals

Catalog Number p-7042808
126.67 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
500 mg
1 g
2 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
62-68-0
C23H32ClNO2
389.964
MFCD00055151
FHIKZROVIDCMJA-UHFFFAOYSA-N
proadifen hydrochloride, proadifen hcl, propyladiphenin, skf-525a hydrochloride, skf 525a, sk&f 525-a, proadifen hydrochloride usan, skf 525-a, skf-525a, hydrochloride
65341
2-(diethylamino)ethyl 2,2-diphenylpentanoate;hydrochloride
CCCC(C1=CC=CC=C1)(C2=CC=CC=C2)C(=O)OCCN(CC)CC.Cl
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
62-68-0
C23H32ClNO2
389.964
MFCD00055151
FHIKZROVIDCMJA-UHFFFAOYSA-N
proadifen hydrochloride, proadifen hcl, propyladiphenin, skf-525a hydrochloride, skf 525a, sk&f 525-a, proadifen hydrochloride usan, skf 525-a, skf-525a, hydrochloride
65341
2-(diethylamino)ethyl 2,2-diphenylpentanoate;hydrochloride
CCCC(C1=CC=CC=C1)(C2=CC=CC=C2)C(=O)OCCN(CC)CC.Cl

Proadifen hydrochloride is an inhibitor of NOS1, AChR, cytochrome P-450, and KIR6. Proadifen hydrochloride bind to the protein moiety of P-450, also reduced the hypoxic response. of Proadifen hydrochloride have good antiarrhythmic effects. SKF-525A substantially reduced the depletion of cardiac norepinephrine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Proadifen hydrochloride is an inhibitor of NOS1, AChR, cytochrome P-450, and KIR6. Proadifen hydrochloride bind to the protein moiety of P-450, also reduced the hypoxic response. of Proadifen hydrochloride have good antiarrhythmic effects. SKF-525A substantially reduced the depletion of cardiac norepinephrine.

Solubility
Soluble in methanol or water

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, heat.
TRUSTED_SUSTAINABILITY
Melting Point 122°C to 123°C
Color White
Quantity 500 mg
Solubility Information Soluble in methanol or water
Formula Weight 389.97
Physical Form Powder
Chemical Name or Material Proadifen hydrochloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'