Learn More
Invitrogen™ ProcartaPlex Human High Sensitivity Basic Kit
Click to view available options
Quantity:
96 Tests
Unit Size:
96 tests
Description
Includes
- Reading Buffer
- Wash Buffer 10x
- Streptavidin-PE
- Universal Assay Buffer
- Detection Antibody Diluent Human
- Amplification Reagent I
- Amplification Reagent II
- PCR 8-Tube Strip
- Adhesive Film
- Greiner Microplate black 96 well
- Quantitate multiple targets in cell/tissue culture supernatants, serum, and plasma
- Explore our broad portfolio of pre-configures panels and combinable single analytes
- For combination of only Simplex Kits an additional Basic Kit is required
- Not intended for human or animal therapeutic or diagnostic use
- Assay diluent, wash buffers, SA-PE, amplification reagents and plates
- This kit does not contain a Multiscreen Filter Plate
Specifications
Specifications
| Product Type | High Sensitivity Simplex Kit |
| Quantity | 96 Tests |
| Species | Human |
| Combinability | Combinable (High Sensitivity Only) |
| Assay Sensitivity | See Certificate of Analysis |
| Format | High Sensitivity Simplex Kit |
| For Use With (Equipment) | Luminex™ Instruments |
| Product Line | ProcartaPlex |