Learn More
Fisherbrand™ Accumet™ Dot-Matrix Printer for AR Series Benchtop Meters: Replacement Ribbon
Product Code. 11864823 Shop All Fisherbrand Products
Product Code. 11864823
Ribbon for Fisherbrand™ Accumet™ Dot-Matrix Printer
Fisherbrand™ Accumet™ Dot-Matrix Printer for accumet AR Series Benchtop Meters: Replacement Ribbon
Description
  • Permanently records results on 2.25 in. (5.7cm) paper with an ink ribbon
  • Prints up to 1.625 lines per second, 28.75 characters per line
  • Operates with large (12,500 lines) or small (3000 lines) roll paper
  • Print speed 38 lines per minute for 24-column
  • Printer communicates via standard RS-232-C interface (also includes RS485 port)
  • Baud rate is 19,200bps
TRUSTED_SUSTAINABILITY
Specifications
Type Ribbon for Dot-Matrix Printer
For Use With (Equipment) Accumet™ Dot-Matrix Printer