Learn More
Fisherbrand™ Accumet™ Dot-Matrix Printer for AR Series Benchtop Meters: Replacement Ribbon
Shop All Fisherbrand ProductsDescription
Fisherbrand™ Accumet™ Dot-Matrix Printer for accumet AR Series Benchtop Meters: Replacement Ribbon
Description
- Permanently records results on 2.25 in. (5.7cm) paper with an ink ribbon
- Prints up to 1.625 lines per second, 28.75 characters per line
- Operates with large (12,500 lines) or small (3000 lines) roll paper
- Print speed 38 lines per minute for 24-column
- Printer communicates via standard RS-232-C interface (also includes RS485 port)
- Baud rate is 19,200bps
Specifications
Specifications
| Type | Ribbon for Dot-Matrix Printer |
| For Use With (Equipment) | Accumet™ Dot-Matrix Printer |