missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Productainer™ BioProcess Container (BPC), 200 L with 4 TopPorts and 1 Bottom Dispense, 3D Single-Use BPC

Using these BPC bags with 3 top and 1 bottom ports, package, store, and transport 50 to 200L volumes of media, buffers, water for injection (WFI), process liquids, and biological products.

1075.00€ - 1255.00€

Specifications

Description Three-dimensional disposable cell culture/bioprocess bag; 100L; w/four top ports, one bottom dispense
Film Description CX3-9 Film
Sterilization Method Gamma Irradiated
Storage Requirements Ambient
Washdown Line No
View More Specs

Products 2
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
15516839 View Documents Thermo Scientific™
SH30652.01
50L 1075.00€
Pack of 5 View alternative products
Save Up to 
»
Estimated Shipment
13-07-2020
Add to basket
 
15526839 View Documents Thermo Scientific™
SH30652.02
100L 1255.00€
Pack of 5 View alternative products
Save Up to 
»
Estimated Shipment
13-07-2020
Add to basket
 
Description

Description

Productainer BPCs are constructed with our Thermo Scientific CX5-14 single web, multi-layered cast film manufactured in a cGMP clean room facility. Our standard Productainers are suitable for a wide array of bioprocess applications. For optimal fit, form and function, Productainers can be customized using the industry's largest component library.

Highlights:

  • Three sizes - 50, 100 and 200L
  • Film optimized for flexibility and durability: superior stress resistance, puncture resistance and physical barrier properties to ensure stability of contents
  • Gamma-irradiation sterilized
  • Squared tube design with polyethylene ports located on the top of the BPC for easy access
  • Space-efficient, ergonomic alternative to rigid walled carboys and steel tanks
  • Customizable via the industry's largest library of qualified components

Recommended for:

  • Harvest - collection and storage of bulk product from bioreactors

  • Storage - sterile storage of media, buffers and process intermediates

  • Separation - Feeding and receiving of sterile liquids from separation system

  • Purification - Buffer feeding to purification system, fraction collection and storage

  • Filling - Sterile reservoir for filling systems

  • Transport and Shipping - Bulk sterile liquids, buffers, media and process intermediates within and between facilities

Specifications

Specifications

Three-dimensional disposable cell culture/bioprocess bag; 100L; w/four top ports, one bottom dispense
Gamma Irradiated
No
CX3-9 Film
Ambient