Learn More
Biochrom Programmable Peltier Heated Cell Holder
Temperature Programmable Peltier Heated Cell Holder with software. Temperature ranges from 20 to 105°C. Used for DNA/RNA denaturing studies.
Brand: Biochrom 80210614
Additional Details : Weight : 0.65000kg
Description
- Applications include the analysis of melting temperatures of oligonucleotides
- Peltier-heated single cell holder for 10 mm pathlength cells
- Compatible with Ultrospec 2100 Pro, 3100, 3300, 4300, 5300 and 6300
- Supplied with cables and the SWIFT-Tm applications software
- Requires Temperature Control Unit
Specifications
Ultrospec 2100 Pro, 3100, 3300, 4300, 5300, 6300 | |
Reusable | |
Software | |
Temperature Control Unit |
Programmable Peltier Heated Cell Holder | |
DNA/RNA Denautring Studies | |
20°C to 105°C | |
20°C to 105°C |