missing translation for 'onlineSavingsMsg'
Learn More

Propargyl Methanesulfonate Ester, 95%, TRC

Product Code. 30411052
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30411052 1 g 1g
30422982 5 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30411052 Supplier Toronto Research Chemicals Supplier No. TRCP7660001G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16156-58-4
Molecular Formula C4 H6 O3 S
Molecular Weight (g/mol) 134.15
IUPAC Name prop-2-ynyl methanesulfonate
SMILES CS(=O)(=O)OCC#C

Specifications

Quantity 1 g
InChI Formula InChI=1S/C4H6O3S/c1-3-4-7-8(2,5)6/h1H,4H2,2H3
Formula Weight 134.0038
Chemical Name or Material Propargyl Methanesulfonate Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.