missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ eBioscience™ Propidium Iodide Staining Solution
Click to view available options
Quantity:
2 mL
Unit Size:
Each
Description
- 5μL (40 ng)/test
- Can be used in FL3 for inviability exclusion, but should be analyzed in FL2 when used as a counterstain for FITC Annexin V
Specifications
Specifications
| Color | Red |
| Detection Method | Fluorescence |
| For Use With (Application) | Viability Assay |
| For Use With (Equipment) | Flow Cytometer |
| Product Type | Staining Solution |
| Dye Type | Cell-Impermeant |
| Emission | 535/617 |
| Product Line | eBioscience |
| Quantity | 2 mL |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction