missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Propyl Methanesulfonate, TRC
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Specifications
Specifications
| Chemical Name or Material | Propyl Methanesulfonate |
| CAS | 1912-31-8 |
| Molecular Formula | C4H10O3S |
| Quantity | 100 mg |
| InChI Formula | InChI=1S/C4H10O3S/c1-3-4-7-8(2,5)6/h3-4H2,1-2H3 |
| SMILES | CCCOS(=O)(=O)C |
| IUPAC Name | propyl methanesulfonate |
| Molecular Weight (g/mol) | 138.18 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction