missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Proteasome Substrate
For cellular and molecular biology applications
31.95€ - 125.00€
Specifications
Color | White |
---|---|
Form | Lyophilized |
For Use With (Application) | Fluorogenic proteasome substrate |
Substrate Type | Proteasome Substrate |
Molecular Weight (g/mol) | 648.79 |
Product Code | Brand | Substrate Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Substrate Quantity | Price | Quantity & Availability | |||||
15406429
|
Thermo Scientific™
J64492.M^ |
1mg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
15496419
|
Thermo Scientific™
J64492.#0 |
5mg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
White | |
Fluorogenic proteasome substrate | |
648.79 | |
-30° to -10°C |
Lyophilized | |
Proteasome Substrate | |
Light Sensitive |
For Research Use Only