missing translation for 'onlineSavingsMsg'
Learn More

Trajan Scientific and Medical™ ProteCo™ PEEK Stainless Steel Unions

Precision engineered for capillary LC. Trajan Scientific and Medical ProteCol™ PEEK Stainless Steel Unions offer complete versatility for connecting capillary tubing.

Marque:  Trajan Scientific and Medical™ 222007

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15279213

  • 123.00€ / 2 pièces


nous consulter 056 260 260
Ajouter au panier

Description

Description

  • Leak free connections
  • Replacement parts such as nuts and ferrules
Spécification

Spécification

PEEK Stainless Steel Unions 0.36mm to 0.36mm
Stainless steel
Size: 0.36 mm to 0.36 mm
Vidéos
Documents