Learn More
Hirschmann™ Protection Rings for Glass Cylinder
Click to view available options
Diameter (Metric) Inner:
14 mm
21 mm
25 mm
29 mm
39 mm
54 mm
Unit Size:
Pack of 5
Specifications
Specifications
| Diameter (Metric) Inner | 14 mm |
| Product Type | Protection Ring |
| Quantity | 5 Pk. |